Gene Transcripts

Gene Symbol
Olfr6
Entrez Gene ID
233670
Gene Name
olfactory receptor 6
Gene Aliases
M50, MOR103-16
Chromosome Location
Chr. 7: 106955984 - 106956934 on Build GRCm38
UniGene ID
Mm.296765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206897.2 NP_996780.2 1 605
GenBank mRNA BC127998.1 - 1 609

View Full Product Details