Gene Transcripts

Gene Symbol
Raver2
Entrez Gene ID
242570
Gene Name
ribonucleoprotein, PTB-binding 2
Gene Aliases
A430091O22Rik
Chromosome Location
Chr. 4: 101069038 - 101152370 on Build GRCm38
UniGene ID
Mm.185636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183024.1 NP_898845.1 3 691
GenBank mRNA AK029114.1 - 1 187
AK039072.1 - 5 579
AK040404.1 - 3 748
BC055329.1 - 3 691 6835616

View Full Product Details