Gene Transcripts

Gene Symbol
Casc4
Entrez Gene ID
319996
Gene Name
cancer susceptibility candidate 4
Gene Aliases
D130060C09Rik, VGFG2573
Chromosome Location
Chr. 2: 121866970 - 121936220 on Build GRCm38
UniGene ID
Mm.292109
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177054.5 NP_796028.2 6, 7 1028
NM_199038.3 NP_950239.2 6, 7 1028
GenBank mRNA AY484583.1 - 6, 7 868
AY484585.1 - 6, 7 868

View Full Product Details