Gene Transcripts

Gene Symbol
Bend6
Entrez Gene ID
320705
Gene Name
BEN domain containing 6
Gene Aliases
B230209C24Rik
Chromosome Location
Chr. 1: 33852052 - 33907767 on Build GRCm38
UniGene ID
Mm.336619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177235.3 NP_796209.2 4 581
GenBank mRNA AK044633.1 - 3 520
AK045534.1 - 3 719
AK134655.1 - 4 724
BC057378.1 - 4 581 5707039

View Full Product Details