Gene Transcripts

Gene Symbol
Rergl
Entrez Gene ID
632971
Gene Name
RERG/RAS-like
Gene Aliases
EG632971
Chromosome Location
Chr. 6: 139493182 - 139501909 on Build GRCm38
UniGene ID
Mm.475610
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128090.1 NP_001121562.1 4 351
GenBank mRNA BC147153.1 - 4 351
BC147154.1 - 4 351 9056193
BC147732.1 - 4 416 9056681
BC147735.1 - 4 426 9056684

View Full Product Details