Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163691.1 | NP_001157163.1 | 10 | 2759 | |
NM_021415.4 | NP_067390.4 | 10 | 2759 | ||
GenBank mRNA | AK147242.1 | - | 10 | 2760 | |
BC138026.1 | - | 10 | 2598 | 8861045 | |
CD350936.1 | - | 1 | 198 |