Gene Transcripts

Gene Symbol
Bcs1l
Entrez Gene ID
66821
Gene Name
BCS1-like (yeast)
Gene Aliases
9130022O19Rik
Chromosome Location
Chr. 1: 74588294 - 74592443 on Build GRCm38
UniGene ID
Mm.358700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK012324.1 - 4 787
AK078925.1 - 4 799
AK079385.1 - 4 795
BC019781.1 - 4 687 4036047

View Full Product Details