Gene Transcripts

Gene Symbol
Mkrn2
Entrez Gene ID
67027
Gene Name
makorin, ring finger protein, 2
Gene Aliases
2610002L04Rik, C81377
Chromosome Location
Chr. 6: 115601938 - 115618670 on Build GRCm38
UniGene ID
Mm.101316
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023290.2 NP_075779.2 3, 4 387
GenBank mRNA AF277171.1 - 3, 4 355
AK009515.1 - 3, 4 396
AK011295.1 - 3, 4 366
AK077926.1 - 3, 4 387
BC025547.1 - 3, 4 406 5320887

View Full Product Details