Gene Transcripts

Gene Symbol
Atp8b3
Entrez Gene ID
67331
Gene Name
ATPase, class I, type 8B, member 3
Gene Aliases
1700042F02Rik, 1700056N23Rik, ATPIK, SAPLT
Chromosome Location
Chr. 10: 80519584 - 80539150 on Build GRCm38
UniGene ID
Mm.52511
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026094.3 NP_080370.2 25, 26 3269
GenBank mRNA AY364445.1 - 25, 26 3269
BC118977.1 - 25, 26 3269 40098417

View Full Product Details