Gene Transcripts

Gene Symbol
Lrriq4
Entrez Gene ID
68307
Gene Name
leucine-rich repeats and IQ motif containing 4
Gene Aliases
4930558O21Rik
Chromosome Location
Chr. 3: 30644506 - 30672431 on Build GRCm38
UniGene ID
Mm.389425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026668.2 NP_080944.2 3, 4 536
GenBank mRNA AK019775.1 - 3, 4 537
BC145806.1 - 3, 4 237 40131109

View Full Product Details