Gene Transcripts

Gene Symbol
Odam
Entrez Gene ID
69592
Gene Name
odontogenic, ameloblast asssociated
Gene Aliases
2310011G06Rik, APIN
Chromosome Location
Chr. 5: 87885025 - 87894174 on Build GRCm38
UniGene ID
Mm.389652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027128.2 NP_081404.2 2 87
GenBank mRNA EF113909.1 - 2 87

View Full Product Details