Gene Transcripts

Gene Symbol
Ttc29
Entrez Gene ID
73301
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
1700031F13Rik
Chromosome Location
Chr. 8: 78213302 - 78394326 on Build GRCm38
UniGene ID
Mm.61243
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183096.3 NP_898919.3 7 839
GenBank mRNA BC048386.1 - 7 839

View Full Product Details