Gene Transcripts

Gene Symbol
Amhr2
Entrez Gene ID
110542
Gene Name
anti-Mullerian hormone type 2 receptor
Gene Aliases
Misiir, Misrii, Mrii
Chromosome Location
Chr. 15: 102445028 - 102454640 on Build GRCm38
UniGene ID
Mm.60331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144547.2 NP_653130.2 11 1792
GenBank mRNA AF503863.1 - 11 1739
BB052908.1 - 1 134
BC119571.1 - 11 1723 40057985
BC119572.2 - 1 73
CJ194934.1 - 2 325

View Full Product Details