Gene Transcripts

Gene Symbol
Aire
Entrez Gene ID
11634
Gene Name
autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
Gene Aliases
Chromosome Location
Chr. 10: 78030022 - 78043610 on Build GRCm38
UniGene ID
Mm.35300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271549.1 NP_001258478.1 10 1204
NM_001271550.1 NP_001258479.1 10 1195
NM_001271551.1 NP_001258480.1 10 1192
NM_009646.2 NP_033776.1 10 1207
GenBank mRNA AF079536.1 - 10 1177
AF128115.1 - 10 1174
AF128116.1 - 10 1165
AF128117.1 - 10 1162
AF128772.1 - 10 1120
AF128773.1 - 10 1120
AJ132243.1 - 10 1120
AJ243821.1 - 10 1120
AK153690.1 - 11 1258
BC103510.1 - 11 1224 40039055
BC103518.1 - 10 1166 40039058
EU625343.3 - 10 1137
JF267399.1 - 10 1137

View Full Product Details