Gene Transcripts
- Gene Symbol
- Aire
- Entrez Gene ID
- 11634
- Gene Name
- autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
- Gene Aliases
- Chromosome Location
- Chr. 10: 78030022 - 78043610 on Build GRCm38
- UniGene ID
- Mm.35300
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271549.1 |
NP_001258478.1
|
6, 7 |
885 |
|
NM_001271552.1 |
NP_001258481.1
|
6, 7 |
885 |
|
NM_001271553.1 |
NP_001258482.1
|
6, 7 |
885 |
|
NM_001271556.1 |
NP_001258485.1
|
6, 7 |
885 |
|
NM_001271557.1 |
NP_001258486.1
|
6, 7 |
885 |
|
NM_009646.2 |
NP_033776.1
|
6, 7 |
885 |
|
GenBank mRNA |
AF079536.1 |
- |
6, 7 |
855 |
|
AF128115.1 |
- |
6, 7 |
855 |
|
AF128118.1 |
- |
6, 7 |
855 |
|
AF128119.1 |
- |
6, 7 |
855 |
|
AF128122.1 |
- |
6, 7 |
855 |
|
AF128123.1 |
- |
6, 7 |
855 |
|
AF128772.1 |
- |
6, 7 |
798 |
|
AF128773.1 |
- |
6, 7 |
798 |
|
AJ132243.1 |
- |
6, 7 |
798 |
|
AJ243821.1 |
- |
6, 7 |
798 |
|
AK153690.1 |
- |
7, 8 |
939 |
|
BC103510.1 |
- |
6, 7 |
847 |
40039055 |
BC103511.1 |
- |
6, 7 |
847 |
40039054 |
BC103518.1 |
- |
6, 7 |
847 |
40039058 |
EU625343.3 |
- |
6, 7 |
815 |
|
JF267399.1 |
- |
6, 7 |
815 |
|
View Full Product Details