Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277944.1 | NP_001264873.1 | 3, 4 | 294 | |
GenBank mRNA | AK146468.1 | - | 2, 3 | 95 | |
BC024697.1 | - | 3, 4 | 294 | 4195301 | |
BG085960.2 | - | 2, 3 | 137 | ||
CB949564.1 | - | 2, 3 | 200 | ||
Z15090.1 | - | 2, 3 | 95 |