Gene Transcripts

Gene Symbol
Bcl7b
Entrez Gene ID
12054
Gene Name
B cell CLL/lymphoma 7B
Gene Aliases
Chromosome Location
Chr. 5: 135168372 - 135181852 on Build GRCm38
UniGene ID
Mm.405834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009745.2 NP_033875.2 5 608
GenBank mRNA AJ011145.1 - 5 488
AK131643.1 - 5 504
AK155076.1 - 5 490
BC011471.1 - 5 585 3660754

View Full Product Details