Gene Transcripts

Gene Symbol
Dmp1
Entrez Gene ID
13406
Gene Name
dentin matrix protein 1
Gene Aliases
AV020965, Dmp, PP
Chromosome Location
Chr. 5: 104197558 - 104214109 on Build GRCm38
UniGene ID
Mm.199008
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016779.2 NP_058059.2 6 727
GenBank mRNA AK132177.1 - 6 728
AK157973.1 - 6 728
BC113753.1 - 6 649 40039844
U65020.1 - 6 733

View Full Product Details