Gene Transcripts

Gene Symbol
Ttc32
Entrez Gene ID
75516
Gene Name
tetratricopeptide repeat domain 32
Gene Aliases
1700013G20Rik
Chromosome Location
Chr. 12: 9029912 - 9045118 on Build GRCm38
UniGene ID
Mm.37581
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029321.2 NP_083597.1 2 295
GenBank mRNA AK005950.1 - 2 291
AK078146.1 - 2 295
AK144458.1 - 2 265
BC119273.1 - 2 279 8734022

View Full Product Details