Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010189.3 | NP_034319.2 | 4 | 749 | |
GenBank mRNA | AK146933.1 | - | 4 | 714 | |
AK152879.1 | - | 4 | 685 | ||
BC003786.1 | - | 4 | 684 | 3485649 | |
D37874.1 | - | 4 | 682 | ||
L17022.1 | - | 3 | 388 |