Gene Transcripts
- Gene Symbol
- Nol6
- Entrez Gene ID
- 230082
- Gene Name
- nucleolar protein family 6 (RNA-associated)
- Gene Aliases
- AA410091, Nrap
- Chromosome Location
- Chr. 4: 41114427 - 41124339 on Build GRCm38
- UniGene ID
- Mm.285797
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_139236.3 |
NP_631982.2
|
2, 3 |
383 |
|
GenBank mRNA |
AF361077.1 |
- |
2, 3 |
332 |
|
AF361078.1 |
- |
2, 3 |
332 |
|
AK053117.1 |
- |
2, 3 |
500 |
|
AK089138.1 |
- |
2, 3 |
303 |
|
AK155818.1 |
- |
2, 3 |
322 |
|
AK166559.1 |
- |
2, 3 |
309 |
|
AK166650.1 |
- |
2, 3 |
309 |
|
AK171163.1 |
- |
2, 3 |
320 |
|
AK171421.1 |
- |
2, 3 |
310 |
|
AK191861.1 |
- |
2, 3 |
239 |
|
AK195706.1 |
- |
2, 3 |
239 |
|
AK197974.1 |
- |
2, 3 |
239 |
|
AK198310.1 |
- |
2, 3 |
239 |
|
AK199838.1 |
- |
2, 3 |
240 |
|
AK202668.1 |
- |
2, 3 |
239 |
|
AK204352.1 |
- |
2, 3 |
242 |
|
AK209500.1 |
- |
2, 3 |
239 |
|
BC059820.1 |
- |
2, 3 |
301 |
6827596 |
BC060189.1 |
- |
2, 3 |
311 |
|
CN459855.1 |
- |
2, 3 |
383 |
|
View Full Product Details