Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025697.3 | NP_079973.1 | 1, 2 | 375 | |
GenBank mRNA | AK018044.1 | - | 1, 2 | 253 | |
AK018153.1 | - | 1, 2 | 376 | ||
AK018290.1 | - | 1, 2 | 225 | ||
AK137685.1 | - | 224 | |||
AK150789.1 | - | 1, 2 | 250 | ||
AK167306.1 | - | 1, 2 | 217 | ||
AK169059.1 | - | 1, 2 | 224 | ||
BC037015.1 | - | 1, 2 | 209 | 5344409 |