Gene Transcripts

Gene Symbol
Thyn1
Entrez Gene ID
77862
Gene Name
thymocyte nuclear protein 1
Gene Aliases
D730042P09Rik, HSPC144, MDS012, MY105, Thy28
Chromosome Location
Chr. 9: 26999659 - 27007334 on Build GRCm38
UniGene ID
Mm.16925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144543.2 NP_653126.1 3 278
GenBank mRNA AB080370.1 - 2 202
AK077639.1 - 3 248
BC016594.1 - 3 232 2631627
CB236103.1 - 2 471

View Full Product Details