Gene Transcripts
- Gene Symbol
- Prkab2
- Entrez Gene ID
- 108097
- Gene Name
- protein kinase, AMP-activated, beta 2 non-catalytic subunit
- Gene Aliases
- 5730553K21Rik, AW049591, BB124140
- Chromosome Location
- Chr. 3: 97658212 - 97673812 on Build GRCm38
- UniGene ID
- Mm.31175
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_182997.2 |
NP_892042.2
|
3, 4 |
493 |
|
GenBank mRNA |
BC060228.1 |
- |
3, 4 |
493 |
|
View Full Product Details