Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007481.3 | NP_031507.1 | 2 | 796 | |
GenBank mRNA | AK006366.1 | - | 1 | 507 | |
AK156960.1 | - | 2 | 781 | ||
BC003478.1 | - | 2 | 758 | 2655889 | |
BC083112.1 | - | 2 | 796 | 5354797 | |
D87903.1 | - | 2 | 738 |