Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007507.2 | NP_031533.2 | 1, 2 | 43 | |
GenBank mRNA | AK011342.1 | - | 1, 2 | 52 | |
BC028438.1 | - | 1, 2 | 43 | 1332765 | |
BC089550.1 | - | 1, 2 | 35 | 30380642 | |
S52977.1 | - | 1, 2 | 26 |