Gene Transcripts

Gene Symbol
Alyref
Entrez Gene ID
21681
Gene Name
Aly/REF export factor
Gene Aliases
ALY, REF1, Refbp1, Thoc4
Chromosome Location
Chr. 11: 120594516 - 120598436 on Build GRCm38
UniGene ID
Mm.1886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011568.1 NP_035698.1 2, 3 387
GenBank mRNA AJ252140.1 - 2, 3 84
BC120588.1 - 2, 3 479 40129511
BC137658.1 - 2, 3 479 8860677
U89876.1 - 2, 3 387

View Full Product Details