Gene Transcripts

Gene Symbol
Acsf2
Entrez Gene ID
264895
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
Chromosome Location
Chr. 11: 94557102 - 94601891 on Build GRCm38
UniGene ID
Mm.386885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153807.2 NP_722502.1 9 1093
GenBank mRNA AK034209.1 - 9 1118
AK153582.1 - 9 1125
AK199766.1 - 4 330
BC018371.1 - 9 1093 4210220
BC063269.1 - 9 1178 4159843

View Full Product Details