Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001082975.1 | NP_001076444.1 | 2 | 109 | |
GenBank mRNA | AK009339.1 | - | 2 | 89 | |
AK049709.1 | - | 2 | 103 | ||
BC058857.1 | - | 1 | 365 | ||
BC087941.1 | - | 2 | 88 | 30303350 |