Gene Transcripts

Gene Symbol
Pnma1
Entrez Gene ID
70481
Gene Name
paraneoplastic antigen MA1
Gene Aliases
5730402C15Rik
Chromosome Location
Chr. 12: 84146131 - 84148489 on Build GRCm38
UniGene ID
Mm.444348
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027438.3 NP_081714.2 1 904
GenBank mRNA AK017476.1 - 1 698
AK028331.1 - 1 842
AK136933.1 - 1 906
BC116687.1 - 1 658 40106117
BC116688.1 - 1 654 40106116

View Full Product Details