Gene Transcripts

Gene Symbol
Vgll3
Entrez Gene ID
73569
Gene Name
vestigial like 3 (Drosophila)
Gene Aliases
1700110N18Rik, 4832416J22, C80713, Vgl-3, Vito-2
Chromosome Location
Chr. 16: 65815017 - 65866368 on Build GRCm38
UniGene ID
Mm.25670
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028572.1 NP_082848.1 3 624
GenBank mRNA AK007165.1 - 2 311
BC042696.1 - 3 549

View Full Product Details