Gene Transcripts

Gene Symbol
Fam83e
Entrez Gene ID
73813
Gene Name
family with sequence similarity 83, member E
Gene Aliases
4930403C10Rik
Chromosome Location
Chr. 7: 45721220 - 45729492 on Build GRCm38
UniGene ID
Mm.322591
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033170.4 NP_001028342.2 5 2258
GenBank mRNA AK015058.1 - 3 584
BC042793.1 - 5 1105

View Full Product Details