Gene Transcripts

Gene Symbol
Iqcf5
Entrez Gene ID
75470
Gene Name
IQ motif containing F5
Gene Aliases
1700007L12Rik
Chromosome Location
Chr. 9: 106514573 - 106516003 on Build GRCm38
UniGene ID
Mm.63799
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029300.1 NP_083576.1 2 384
GenBank mRNA AK005734.1 - 2 385
BC125546.1 - 2 343 40130061

View Full Product Details