Gene Transcripts

Gene Symbol
Plvap
Entrez Gene ID
84094
Gene Name
plasmalemma vesicle associated protein
Gene Aliases
MECA32, Pv1
Chromosome Location
Chr. 8: 71497753 - 71511804 on Build GRCm38
UniGene ID
Mm.260768
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032398.2 NP_115774.2 1 320
GenBank mRNA AF348403.1 - 1 291
AF369900.1 - 1 315
BC013517.1 - 1 323 4235957

View Full Product Details