Gene Transcripts

Gene Symbol
Vof21
Entrez Gene ID
259228
Gene Name
ischemia related factor vof-21
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB089205.1 - 1 1549

View Full Product Details