Gene Transcripts

Gene Symbol
Tatdn3
Entrez Gene ID
289378
Gene Name
TatD DNase domain containing 3
Gene Aliases
Chromosome Location
Chr. 13: 109655292 - 109669573 on Build Rnor_6.0
UniGene ID
Rn.12992
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105987.1 NP_001099457.1 9 812

View Full Product Details