Gene Transcripts

Gene Symbol
Dync1h1
Entrez Gene ID
29489
Gene Name
dynein cytoplasmic 1 heavy chain 1
Gene Aliases
Dnch1, Dnchc1
Chromosome Location
Chr. 6: 134958854 - 135085769 on Build Rnor_6.0
UniGene ID
Rn.11027
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019226.3 NP_062099.3 24 5098
GenBank mRNA L08505.1 - 24 5098

View Full Product Details