Gene Transcripts

Gene Symbol
Mrps9
Entrez Gene ID
301371
Gene Name
mitochondrial ribosomal protein S9
Gene Aliases
Chromosome Location
Chr. 9: 49647246 - 49704828 on Build Rnor_6.0
UniGene ID
Rn.13913
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100549.1 NP_001094019.1 9 861
XM_008767019.1 XP_008765241.1 9 872
GenBank mRNA BC088242.1 - 8 673
BC158704.1 - 9 829 9030006

View Full Product Details