Gene Transcripts

Gene Symbol
Rnf20
Entrez Gene ID
313216
Gene Name
ring finger protein 20, E3 ubiquitin protein ligase
Gene Aliases
Chromosome Location
Chr. 5: 64893001 - 64920397 on Build Rnor_6.0
UniGene ID
Rn.15969
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107929.2 NP_001101399.2 16 2725
XM_006238154.2 XP_006238216.1 17 2650
XM_006238155.2 XP_006238217.1 17 2569

View Full Product Details