Gene Transcripts

Gene Symbol
Snx16
Entrez Gene ID
64088
Gene Name
sorting nexin 16
Gene Aliases
Chromosome Location
Chr. 2: 93518796 - 93541076 on Build Rnor_6.0
UniGene ID
Rn.64585
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022289.3 NP_071625.2 7 1186
GenBank mRNA AF305780.1 - 7 1172
BC061554.1 - 7 1190 6890629

View Full Product Details