Gene Transcripts

Gene Symbol
Bpnt1
Entrez Gene ID
64473
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
Sal3
Chromosome Location
Chr. 13: 103268046 - 103292848 on Build Rnor_6.0
UniGene ID
Rn.8453
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_171990.2 NP_741987.1 9 934
XM_006250429.2 XP_006250491.1 10 1021
XM_006250430.2 XP_006250492.1 10 1076
XM_006250431.2 XP_006250493.1 9 1030
GenBank mRNA AJ000347.1 - 9 885
BC085692.1 - 9 934 7190529
FQ214355.1 - 10 1002

View Full Product Details