Gene Transcripts

Gene Symbol
LOC171573
Entrez Gene ID
171573
Gene Name
spleen protein 1 precursor
Gene Aliases
Sslp1
Chromosome Location
Chr. 8: 41442898 - 41445722 on Build Rnor_6.0
UniGene ID
Rn.228922
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138537.2 NP_612546.1 2, 3 192
GenBank mRNA AF198442.1 - 2, 3 174
CB695439.1 - 2, 3 213
FQ228610.1 - 2, 3 212

View Full Product Details