Gene Transcripts

Gene Symbol
Arrdc4
Entrez Gene ID
293019
Gene Name
arrestin domain containing 4
Gene Aliases
Ab1-209, LRRG00117
Chromosome Location
Chr. 1: 129766097 - 129780151 on Build Rnor_6.0
UniGene ID
Rn.14864
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047853.1 NP_001041318.1 2 58
GenBank mRNA AY325151.1 - 2 58
AY539868.1 - 1 18

View Full Product Details