Gene Transcripts

Gene Symbol
Stac2
Entrez Gene ID
363674
Gene Name
SH3 and cysteine rich domain 2
Gene Aliases
Chromosome Location
Chr. 10: 85988367 - 86004209 on Build Rnor_6.0
UniGene ID
Rn.153636
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108834.1 NP_001102304.1 3 893
XM_006247383.2 XP_006247445.1 5 973
GenBank mRNA AM903074.1 - 5 607

View Full Product Details