Gene Transcripts
- Gene Symbol
- Mir17hg
- Entrez Gene ID
- 75957
- Gene Name
- Mir17 host gene (non-protein coding)
- Gene Aliases
- 5033413D16Rik, Mirhg1, OTTMUSG00000033261
- Chromosome Location
- Chr. 14: 115044390 - 115046728 on Build GRCm38
- UniGene ID
- Mm.34859
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029382.1 |
|
1 |
1186 |
|
GenBank mRNA |
AK017164.1 |
- |
1 |
50 |
|
AK053349.1 |
- |
1 |
1079 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE332918.1 |
- |
1 |
385 |
|
Chr. 14 - 115045191 - 115045686 [+] |
GenBankmRNA |
BF019638.1 |
- |
1 |
255 |
|
Chr. 14 - 115045321 - 115045741 [+] |
GenBankmRNA |
AA174312.1 |
- |
1 |
544 |
|
Chr. 14 - 115045032 - 115045587 [+] |
GenBankmRNA |
CD548669.1 |
- |
1 |
549 |
|
Chr. 14 - 115045027 - 115045620 [+] |
View Full Product Details