Gene Transcripts
- Gene Symbol
- Asb7
- Entrez Gene ID
- 117589
- Gene Name
- ankyrin repeat and SOCS box-containing 7
- Gene Aliases
- AI449039, Asb-7, D030055C23Rik
- Chromosome Location
- Chr. 7: 66644565 - 66689650 on Build GRCm38
- UniGene ID
- Mm.34785
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003961.1 |
|
2, 3 |
569 |
|
GenBank mRNA |
AK051017.1 |
- |
2, 3 |
482 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB655391.1 |
- |
2, 3 |
479 |
|
Chr. 7 - 66681704 - 66689472 [-] |
View Full Product Details