Gene Transcripts
- Gene Symbol
- Mir423
- Entrez Gene ID
- 751519
- Gene Name
- microRNA 423
- Gene Aliases
- Mirn423, mir-423, mmu-mir-423
- Chromosome Location
- Chr. 11: 77078064 - 77078172 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030756.1 |
|
1 |
53 |
|
- Gene Symbol
- Ccdc55
- Entrez Gene ID
- 237859
- Gene Name
- coiled-coil domain containing 55
- Gene Aliases
- 8030451K07, AI851076, AL022898, NSpr70, NSrp70, Nsrp1
- Chromosome Location
- Chr. 11: 77044292 - 77078437 on Build GRCm38
- UniGene ID
- Mm.156195
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038056.1 |
- |
1 |
318 |
|
View Full Product Details