Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003226.1 | 8 | 984 | ||
NR_003227.1 | 7 | 811 | |||
NR_003228.1 | 8 | 950 | |||
GenBank mRNA | AF329691.1 | - | 7 | 735 | |
AF329692.1 | - | 2 | 286 | ||
AK056488.1 | - | 8 | 955 | ||
AK091293.1 | - | 8 | 947 | ||
AK097044.1 | - | 7 | 798 | ||
AK126911.1 | - | 6 | 775 | ||
AK128352.1 | - | 1 | 650 | ||
AL832984.1 | - | 2 | 208 | ||
BC021245.2 | - | 6 | 557 |