Gene Transcripts
- Gene Symbol
- HSD52
- Entrez Gene ID
- 729467
- Gene Name
- uncharacterized LOC729467
- Gene Aliases
- Chromosome Location
- Chr. 1: 59131936 - 59146807 on Build GRCh38
- UniGene ID
- Hs.662291
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027120.1 |
|
3 |
405 |
|
GenBank mRNA |
DB453888.1 |
- |
3 |
405 |
|
DQ141102.1 |
- |
1 |
25 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY024377.1 |
- |
3 |
429 |
|
Chr. 1 - 59132534 - 59146830 [-] |
GenBankmRNA |
AA397748.1 |
- |
3 |
325 |
|
Chr. 1 - 59132466 - 59146727 [-] |
GenBankmRNA |
HY013413.1 |
- |
3 |
418 |
|
Chr. 1 - 59132484 - 59146820 [-] |
View Full Product Details