Gene Transcripts

Gene Symbol
SDK1
Entrez Gene ID
221935
Gene Name
sidekick cell adhesion molecule 1
Gene Aliases
Chromosome Location
Chr. 7: 3301448 - 4269000 on Build GRCh38
UniGene ID
Hs.653013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079653.1 NP_001073121.1 1 39
GenBank mRNA AL833354.1 - 1 36
DB065853.1 - 1 35
DB454012.1 - 1 39

View Full Product Details